![Doxepin impurity standard British Pharmacopoeia (BP) Reference Standard](/deepweb/assets/sigmaaldrich/product/structures/218/125/6d8e0f2d-385b-46cd-b40c-e7336168f66a/640/6d8e0f2d-385b-46cd-b40c-e7336168f66a.png)
Doxepin impurity standard
![Doxepin impurity standard British Pharmacopoeia (BP) Reference Standard](/deepweb/assets/sigmaaldrich/product/structures/218/125/6d8e0f2d-385b-46cd-b40c-e7336168f66a/640/6d8e0f2d-385b-46cd-b40c-e7336168f66a.png)
Synonym(s):
Doxepin hydrochloride, 11-(3-Dimethylaminopropylidene)-6,11-dihydrodibenz[b,e]oxepine hydrochloride
Empirical Formula (Hill Notation):
C19H21NO · HCl
CAS Number:
Molecular Weight:
315.84
Products
Formula Weight
Manufacturer
Physical Form
Format
Application
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing