DMT-2′O-MOE-rA(bz) Phosphoramidite
Synonym(s):
5′-Dimethoxytrityl-N6-Benzoyl-3′-[(2-cyanoethyl)- (N,N-diisopropyl)]-phosphoramidite- 2′-O-(2-methoxyethyl)adenosine
Linear Formula:
C50H58N7O9P
CAS Number:
Molecular Weight:
932.01
Products
Product Category
Brand
Formula Weight
Physical Form
Quality Segment
Showing 1-1 of 1
Compare
Product Number
Description
Pricing