![D-Mannose 6-phosphate sodium salt ≥98% (HPLC)](/deepweb/assets/sigmaaldrich/product/structures/388/044/8f925bd3-4003-4453-a9d8-18d2a14ecb06/640/8f925bd3-4003-4453-a9d8-18d2a14ecb06.png)
D-Mannose 6-phosphate sodium salt
![D-Mannose 6-phosphate sodium salt ≥98% (HPLC)](/deepweb/assets/sigmaaldrich/product/structures/388/044/8f925bd3-4003-4453-a9d8-18d2a14ecb06/640/8f925bd3-4003-4453-a9d8-18d2a14ecb06.png)
Linear Formula:
C6H12O9PNa
CAS Number:
Molecular Weight:
282.12
Products
Product Category
Brand
Biological Source
Color
Formula Weight
Physical Form
Purity
Quality Segment
Optical Isomer
Technique
Showing 1-1 of 1
Compare
Product Number
Description
Pricing