![DL-Methionine European Pharmacopoeia (EP) Reference Standard](/deepweb/assets/sigmaaldrich/product/structures/339/066/d347df59-b15b-448c-9b8a-5558fb145250/640/d347df59-b15b-448c-9b8a-5558fb145250.png)
DL-Methionine
![DL-Methionine European Pharmacopoeia (EP) Reference Standard](/deepweb/assets/sigmaaldrich/product/structures/339/066/d347df59-b15b-448c-9b8a-5558fb145250/640/d347df59-b15b-448c-9b8a-5558fb145250.png)
Synonym(s):
(±)-2-Amino-4-(methylmercapto)butyric acid, DL-2-Amino-4-(methylthio)butanoic acid
Linear Formula:
CH3SCH2CH2CH(NH2)COOH
CAS Number:
Molecular Weight:
149.21
Products
Product Category
Brand
Biological Source
Color
Formula Weight
Manufacturer
Melting Point (°C)
Physical Form
Product Line
Purity
Agency
Format
Quality Segment
Application
Organoleptic Class
Amino Acid
API Family
Reference Material Type
Special Grade
Technique
Showing 1-5 of 5
Compare
Product Number
Description
Pricing