Skip to Content
Merck
DL-Lactonitrile ≥97.0% (T)

DL-Lactonitrile

DL-Lactonitrile ≥97.0% (T)
Synonym(s):
2-Hydroxypropionitrile, Acetaldehyde cyanohydrin
Linear Formula:
CH3CH(OH)CN
CAS Number:
Molecular Weight:
71.08

Products

Product Category
Brand
Boiling Point (°C)
Formula Weight
Physical Form
Purity
Quality Segment
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing