![(±)-Epinephrine hydrochloride solution 1 mg/mL in methanol ((as free base)), certified reference material, ampule of 1 mL, Cerilliant®](/deepweb/assets/sigmaaldrich/product/structures/166/437/ff63fb48-cdc0-4ce6-8558-ef65379396d4/640/ff63fb48-cdc0-4ce6-8558-ef65379396d4.png)
(±)-Epinephrine hydrochloride solution
![(±)-Epinephrine hydrochloride solution 1 mg/mL in methanol ((as free base)), certified reference material, ampule of 1 mL, Cerilliant®](/deepweb/assets/sigmaaldrich/product/structures/166/437/ff63fb48-cdc0-4ce6-8558-ef65379396d4/640/ff63fb48-cdc0-4ce6-8558-ef65379396d4.png)
Synonym(s):
(±)-Epinephrine hydrochloride, (±)-Adrenalin, 4-(1-Hydroxy-2-[methylamino]ethyl)-1,2-benzenediol hydrochloride
Empirical Formula (Hill Notation):
C9H13NO3 · HCl
CAS Number:
Molecular Weight:
219.67
Products
Brand
Formula Weight
Manufacturer
Format
Quality Segment
Package Size
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing