![Dipyrrolidino(N-succinimidyloxy)carbenium hexafluorophosphate ≥98.0% (CHN)](/deepweb/assets/sigmaaldrich/product/structures/304/938/7327661f-35cc-4b5a-8a3b-ceaa57fa32b7/640/7327661f-35cc-4b5a-8a3b-ceaa57fa32b7.png)
Dipyrrolidino(N-succinimidyloxy)carbenium hexafluorophosphate
![Dipyrrolidino(N-succinimidyloxy)carbenium hexafluorophosphate ≥98.0% (CHN)](/deepweb/assets/sigmaaldrich/product/structures/304/938/7327661f-35cc-4b5a-8a3b-ceaa57fa32b7/640/7327661f-35cc-4b5a-8a3b-ceaa57fa32b7.png)
Synonym(s):
N,N,N′,N′-Bis(tetramethylene)-O-(N-succinimidyl)uronium hexafluorophosphate, O-(N-Succinimidyl)-N,N,N′,N′-bis(tetramethylene)uronium hexafluorophosphate, HSPyU
Empirical Formula (Hill Notation):
C13H20F6N3O3P
CAS Number:
Molecular Weight:
411.28
Products
Brand
Formula Weight
Melting Point (°C)
Physical Form
Purity
Quality Segment
Application
Reaction Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing