Skip to Content
Merck
Diphenyl N-cyanocarbonimidate 97%

Diphenyl N-cyanocarbonimidate

Diphenyl N-cyanocarbonimidate 97%
Linear Formula:
NCN=C(OC6H5)2
CAS Number:
Molecular Weight:
238.24

Products

Product Category
Brand
Formula Weight
Gene Alias
Melting Point (°C)
Physical Form
Purity
Quality Segment
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing