Dimethylammonium dimethylcarbamate
Synonym(s):
N-Methylmethanamine dimethylcarbamate, Dimcarb, Dimethylamine carbon dioxide complex
Linear Formula:
(CH3)2NH · (CH3)2NCOOH
CAS Number:
Molecular Weight:
134.18
Products
Product Category
Brand
Boiling Point (°C)
Formula Weight
Physical Form
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing