Skip to Content
Merck
Dimethyl 5-hydroxyisophthalate 98%

Dimethyl 5-hydroxyisophthalate

Dimethyl 5-hydroxyisophthalate 98%
Synonym(s):
Dimethyl 5-hydroxybenzene-1,3-dicarboxylate
Linear Formula:
HOC6H3-1,3-(CO2CH3)2
CAS Number:
Molecular Weight:
210.18
EC Number:

Products

Product Category
Brand
Formula Weight
Melting Point (°C)
Physical Form
Purity
Functional Group
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing