Diisopropyl malonate
Linear Formula:
(CH3)2CHO2CCH2CO2CH(CH3)2
Products
Product Category
Brand
Boiling Point (°C)
Formula Weight
Physical Form
Product Line
Purity
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing