Skip to Content
Merck
Diglycolic anhydride technical grade, 90%

Diglycolic anhydride

Diglycolic anhydride technical grade, 90%
Synonym(s):
1,4-Dioxane-2,6-dione, Diglycolic anhydride
Empirical Formula (Hill Notation):
C4H4O4
CAS Number:
Molecular Weight:
116.07

Products

Brand
Boiling Point (°C)
Formula Weight
Melting Point (°C)
Physical Form
Purity
Quality Segment
Markush Class
Markush Group
Monomer Type
Special Grade
Showing 1-2 of 2
Compare
Product Number
Description
Pricing