![Diethyl (Boc-amino)malonate 97%](/deepweb/assets/sigmaaldrich/product/structures/230/821/9d204728-cb34-42ce-9c9b-cdbc93b4e672/640/9d204728-cb34-42ce-9c9b-cdbc93b4e672.png)
Diethyl (Boc-amino)malonate
![Diethyl (Boc-amino)malonate 97%](/deepweb/assets/sigmaaldrich/product/structures/230/821/9d204728-cb34-42ce-9c9b-cdbc93b4e672/640/9d204728-cb34-42ce-9c9b-cdbc93b4e672.png)
Synonym(s):
(Boc-amino)malonic acid diethyl ester, Diethyl 2-[N-(tert-butoxycarbonyl)amino]malonate
Linear Formula:
(CH3)3COCONHCH(CO2CH2CH3)2
CAS Number:
Molecular Weight:
275.30
Products
Product Category
Brand
Boiling Point (°C)
Formula Weight
Purity
Quality Segment
Application
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing