![Diazoxide European Pharmacopoeia (EP) Reference Standard](/deepweb/assets/sigmaaldrich/product/structures/220/160/6ad4e840-0ece-4382-8ef4-c4f2d75c4279/640/6ad4e840-0ece-4382-8ef4-c4f2d75c4279.png)
Diazoxide
![Diazoxide European Pharmacopoeia (EP) Reference Standard](/deepweb/assets/sigmaaldrich/product/structures/220/160/6ad4e840-0ece-4382-8ef4-c4f2d75c4279/640/6ad4e840-0ece-4382-8ef4-c4f2d75c4279.png)
Synonym(s):
7-Chloro-3-methyl-2H-1,2,4-benzothiadiazine 1,1-dioxide
Empirical Formula (Hill Notation):
C8H7ClN2O2S
CAS Number:
Molecular Weight:
230.67
Products
Product Category
Brand
Formula Weight
Gene Alias
Manufacturer
Physical Form
Format
Quality Segment
Application
API Family
Reference Material Type
Special Grade
Showing 1-3 of 3