![Dexbrompheniramine maleate United States Pharmacopeia (USP) Reference Standard](/deepweb/assets/sigmaaldrich/product/structures/142/127/8d6bb0bd-b628-46ae-95c5-d70fb37ed1eb/640/8d6bb0bd-b628-46ae-95c5-d70fb37ed1eb.png)
Dexbrompheniramine maleate
![Dexbrompheniramine maleate United States Pharmacopeia (USP) Reference Standard](/deepweb/assets/sigmaaldrich/product/structures/142/127/8d6bb0bd-b628-46ae-95c5-d70fb37ed1eb/640/8d6bb0bd-b628-46ae-95c5-d70fb37ed1eb.png)
Synonym(s):
(+)-Brompheniramine maleate salt, Dexbrompheniramine maleate salt
Empirical Formula (Hill Notation):
C16H19BrN2 · C4H4O4
CAS Number:
Molecular Weight:
435.31
Products
Formula Weight
Gene Alias
Manufacturer
Format
Application
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing