Dehydroacetic acid sodium salt
Synonym(s):
Dehydroacetic acid sodium salt
Empirical Formula (Hill Notation):
C8H7NaO4
CAS Number:
Molecular Weight:
190.13
Products
Brand
Formula Weight
Physical Form
Quality Segment
Showing 1-1 of 1
Compare
Product Number
Description
Pricing