Skip to Content
Merck
(+)-α-Methyl-4-carboxyphenylglycine solid

(+)-α-Methyl-4-carboxyphenylglycine

(+)-α-Methyl-4-carboxyphenylglycine solid
Synonym(s):
(+)-MCPG, S(+)-α-Amino-4-carboxy-α-methyl-benzeneacetic acid
Empirical Formula (Hill Notation):
C10H11NO4
CAS Number:
Molecular Weight:
209.20

Products

Product Category
Brand
Formula Weight
Gene Alias
Physical Form
Showing 1-1 of 1
Compare
Product Number
Description
Pricing