Skip to Content
Merck
D-α-Hydroxyglutaric acid disodium salt ≥98.0% (GC)

D-α-Hydroxyglutaric acid disodium salt

D-α-Hydroxyglutaric acid disodium salt ≥98.0% (GC)
Synonym(s):
(R)-2-Hydroxypentanedioic acid disodium salt, Disodium (R)-2-hydroxyglutarate
Linear Formula:
C5H6O5Na2
CAS Number:
Molecular Weight:
192.08

Products

Product Category
Brand
Color
Formula Weight
Physical Form
Purity
Quality Segment
Application
Markush Class
Markush Group
Optical Isomer