Skip to Content
Merck
Cyclopentadienyltitanium(IV) trichloride 97%

Cyclopentadienyltitanium(IV) trichloride

Cyclopentadienyltitanium(IV) trichloride 97%
Empirical Formula (Hill Notation):
C5H5Cl3Ti
CAS Number:
Molecular Weight:
219.32

Products

Brand
Formula Weight
Melting Point (°C)
Physical Form
Reagent Type
Core
Showing 1-1 of 1
Compare
Product Number
Description
Pricing