Skip to Content
Merck
Cyclohexanone-2,2,6,6-d4 98 atom % D

Cyclohexanone-2,2,6,6-d4

Cyclohexanone-2,2,6,6-d4 98 atom % D
Empirical Formula (Hill Notation):
C6D4H6O
CAS Number:
Molecular Weight:
102.17

Products

Product Category
Brand
Boiling Point (°C)
Formula Weight
Melting Point (°C)
Isotope
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing