Crotonic acid
Synonym(s):
trans-2-Butenoic acid, Crotonic acid
Linear Formula:
CH3CH=CHCOOH
CAS Number:
Molecular Weight:
86.09
Products
Product Category
Brand
Boiling Point (°C)
Formula Weight
Melting Point (°C)
Physical Form
Purity
Quality Segment
Markush Class
Markush Group
Showing 1-2 of 2
Compare
Product Number
Description
Pricing