Color Related Compound 005
Synonym(s):
3-Hydroxy-2-naphthoic acid, β-Hydroxynaphthoic acid
Linear Formula:
HOC10H6CO2H
CAS Number:
Molecular Weight:
188.18
Products
Formula Weight
Manufacturer
Melting Point (°C)
Format
Application
Markush Class
Markush Group
API Family
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing