Skip to Content
Merck
Clear Fluid Thioglycollate Medium tube of, ready-to-use, tube volume 15 mL , filling volume

Clear Fluid Thioglycollate Medium

Clear Fluid Thioglycollate Medium tube of, ready-to-use, tube volume 15 mL , filling volume

Products

Product Category
Brand
Physical Form
Sterilization Compatibility
Agency
Capacity (mL)
Volume (ml)
Compatibility
Quality Segment
Closure Type
Application
Microorganism Suitability
Package Size
Sterility
Sterilization Method