Skip to Content
Merck
Clarithromycin Impurity D certified reference material, pharmaceutical secondary standard

Clarithromycin Impurity D

Clarithromycin Impurity D certified reference material, pharmaceutical secondary standard
Synonym(s):
N-Demethylclarithromycin, N-Desmethyl clarithromycin
Empirical Formula (Hill Notation):
C37H67NO13
CAS Number:
Molecular Weight:
733.93

Products

Brand
Formula Weight
Manufacturer
Agency
Format
Quality Segment
Application
Package Size
API Family
Reference Material Type
Special Grade