![cis-11-Vaccenic acid methyl ester certified reference material, 10 mg/mL in heptane, ampule of 1 mL](/deepweb/assets/sigmaaldrich/product/images/208/280/53a73689-a9ce-4352-97fd-dac81add42d3/640/53a73689-a9ce-4352-97fd-dac81add42d3.jpg)
cis-11-Vaccenic acid methyl ester
![cis-11-Vaccenic acid methyl ester certified reference material, 10 mg/mL in heptane, ampule of 1 mL](/deepweb/assets/sigmaaldrich/product/images/208/280/53a73689-a9ce-4352-97fd-dac81add42d3/640/53a73689-a9ce-4352-97fd-dac81add42d3.jpg)
Synonym(s):
cis-11-Octadecenoic methyl ester, cis-Vaccenic acid methyl ester
Linear Formula:
CH3(CH2)5CH=CH(CH2)9CO2CH3
CAS Number:
Molecular Weight:
296.49
Products
Product Category
Brand
Formula Weight
Product Line
Format
Quality Segment
Functional Group
Application
Package Size
Reference Material Type
Special Grade
Technique
Showing 1-1 of 1
Compare
Product Number
Description
Pricing