Skip to Content
Merck
Cinoxacin

Cinoxacin

Cinoxacin
Synonym(s):
1-Ethyl-1,4-dihydro-4-oxo-[1,3]dioxolo[4,5-g]cinnoline-3-carboxylic acid
Empirical Formula (Hill Notation):
C12H10N2O5
CAS Number:
Molecular Weight:
262.22
EC Number:

Products

Product Category
Brand
Formula Weight
Physical Form
Activity Spectrum
Mode of Action
Showing 1-1 of 1
Compare
Product Number
Description
Pricing