
Chlorosulfuron

Synonym(s):
1-(2-Chlorophenylsulfonyl)-3-(4-methoxy-6-methyl-1,3,5-triazin-2-yl)-urea, Chlorsulfuron
Empirical Formula (Hill Notation):
C12H12ClN5O4S
Products
Product Category
Formula Weight
Product Line
Format
Quality Segment
Application
Reference Material Type
Special Grade
Technique
Showing 1-1 of 1
Compare
Product Number
Description
Pricing