Chlormadinone acetate
Synonym(s):
17α-Acetoxy-6-chloro-4,6-pregnadiene-3,20-dione, 6-Chloro-17α-hydroxy-4,6-pregnadiene-3,20-dione 17-acetate, 6-Chloro-6-dehydro-17α-acetoxyprogesterone, Gestafortin, Matrol, Menstridyl
Empirical Formula (Hill Notation):
C23H29ClO4
CAS Number:
Molecular Weight:
404.93
Products
Brand
Formula Weight
Gene Alias
Manufacturer
Physical Form
Purity
Format
Application
API Family
Reference Material Type
Special Grade
Showing 1-2 of 2
Compare
Product Number
Description
Pricing