![Cefazolin sodium British Pharmacopoeia (BP) Reference Standard](/deepweb/assets/sigmaaldrich/product/structures/237/174/e8a4217d-fff9-418b-84a0-8e67bc22aabf/640/e8a4217d-fff9-418b-84a0-8e67bc22aabf.png)
Cefazolin sodium
![Cefazolin sodium British Pharmacopoeia (BP) Reference Standard](/deepweb/assets/sigmaaldrich/product/structures/237/174/e8a4217d-fff9-418b-84a0-8e67bc22aabf/640/e8a4217d-fff9-418b-84a0-8e67bc22aabf.png)
Synonym(s):
Cefazolin sodium salt
Empirical Formula (Hill Notation):
C14H13N8NaO4S3
Products
Formula Weight
Manufacturer
Physical Form
Format
Application
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing