Skip to Content
Merck
Caprylic Acid (Octanoic Acid) Pharmaceutical Secondary Standard; Certified Reference Material

Caprylic acid (Octanoic acid)

Caprylic Acid (Octanoic Acid) Pharmaceutical Secondary Standard; Certified Reference Material
Synonym(s):
Caprylic acid (Octanoic acid), Octanoic acid, Acid C8, Caprylic acid
Linear Formula:
CH3(CH2)6COOH
CAS Number:
Molecular Weight:
144.21
EC Number:

Products

Product Category
Brand
Boiling Point (°C)
Formula Weight
Melting Point (°C)
Physical Form
Product Line
Agency
Format
Quality Segment
Application
Markush Class
Markush Group
API Family
Reference Material Type
Special Grade
Technique