Skip to Content
Merck
Calyculin A from Discodermia calyx ≥90% (HPLC), solid

Calyculin A

Calyculin A from Discodermia calyx ≥90% (HPLC), solid
Synonym(s):
Calyculin A, Discodermia calyx
Empirical Formula (Hill Notation):
C50H81N4O15P
CAS Number:
Molecular Weight:
1009.17

Products

Product Category
Brand
Biological Source
Color
Formula Weight
Physical Form
Purity
Research Area
Quality Segment