Calmidazolium chloride
Synonym(s):
1-[Bis(4-chlorophenyl)methyl]-3-[2,4-dichloro-β-(2,4-dichlorobenzyloxy)phenethyl]imidazolium chloride, 1-[bis(4-Chlorophenyl)methyl]-3-[2-(2,4-dichlorophenyl)-2-(2,4-dichlorobenzyloxy)ethyl]-1H-imidazolium chloride, R 24571
Empirical Formula (Hill Notation):
C31H23Cl7N2O
CAS Number:
Molecular Weight:
687.70
Products
Product Category
Brand
Color
Formula Weight
Physical Form
Research Area
Quality Segment
Showing 1-1 of 1
Compare
Product Number
Description
Pricing