![Brominated Wang resin 100-200 mesh, extent of labeling: 0.5-1.5 mmol/g loading, 1 % cross-linked](/deepweb/assets/sigmaaldrich/product/structures/331/251/61c86643-47d8-46d5-97d2-490f5b1c5a65/640/61c86643-47d8-46d5-97d2-490f5b1c5a65.png)
Brominated Wang resin
![Brominated Wang resin 100-200 mesh, extent of labeling: 0.5-1.5 mmol/g loading, 1 % cross-linked](/deepweb/assets/sigmaaldrich/product/structures/331/251/61c86643-47d8-46d5-97d2-490f5b1c5a65/640/61c86643-47d8-46d5-97d2-490f5b1c5a65.png)
Synonym(s):
α-Bromopropiophenone, polymer-bound, 2-Bromopropionylpolystyrene
Products
Product Category
Brand
Quality Segment
Application
Cleavage Condition
Linker
Chain Type
Reaction Type
Showing 1-1 of 1
Compare
Product Number
Description
Pricing