![Boeravinone B phyproof® Reference Substance](/deepweb/assets/sigmaaldrich/product/structures/304/908/ebf8a455-5ecc-4a2e-89cc-f30cb11cc03e/640/ebf8a455-5ecc-4a2e-89cc-f30cb11cc03e.png)
Boeravinone B
![Boeravinone B phyproof® Reference Substance](/deepweb/assets/sigmaaldrich/product/structures/304/908/ebf8a455-5ecc-4a2e-89cc-f30cb11cc03e/640/ebf8a455-5ecc-4a2e-89cc-f30cb11cc03e.png)
Synonym(s):
6,9,11-Trihydroxy-10-methyl-[1]benzopyrano[3,4-b][1]benzopyran-12(6H)-one
Empirical Formula (Hill Notation):
C17H12O6
CAS Number:
Molecular Weight:
312.27
Products
Formula Weight
Manufacturer
Physical Form
Product Line
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing