Skip to Content
Merck
Boc-Cys(Acm)-OH Novabiochem®

Boc-Cys(Acm)-OH

Boc-Cys(Acm)-OH Novabiochem®
Synonym(s):
Boc-Cys(Acm)-OH
Empirical Formula (Hill Notation):
C11H20N2O5S
CAS Number:
Molecular Weight:
292.35

Products

Product Category
Brand
Formula Weight
Physical Form
Product Line
Quality Segment
Functional Group
Application
Optical Isomer
Amino Acid
Reaction Type
Showing 1-1 of 1
Compare
Product Number
Description
Pricing