
Boc-Asp-OH

Synonym(s):
Boc-Asp-OH
Empirical Formula (Hill Notation):
C9H15NO6
CAS Number:
Molecular Weight:
233.22
Products
Product Category
Brand
Formula Weight
Physical Form
Product Line
Quality Segment
Functional Group
Application
Optical Isomer
Amino Acid
Reaction Type
Showing 1-1 of 1
Compare
Product Number
Description
Pricing