Skip to Content
Merck
Boc-2-Nal-OH ≥97.0% (HPLC)

Boc-2-Nal-OH

Boc-2-Nal-OH ≥97.0% (HPLC)
Synonym(s):
Boc-3-(2-naphthyl)-L-alanine
Empirical Formula (Hill Notation):
C18H21NO4
CAS Number:
Molecular Weight:
315.36

Products

Brand
Formula Weight
Melting Point (°C)
Purity
Application
Markush Class
Markush Group
Optical Isomer
Amino Acid
Amino Acid Type
Reaction Type
Showing 1-1 of 1
Compare
Product Number
Description
Pricing