Skip to Content
Merck
Bisoprolol hemifumarate salt ≥98% (HPLC), solid

Bisoprolol hemifumarate salt

Bisoprolol hemifumarate salt ≥98% (HPLC), solid
Synonym(s):
1-[4-[[2-(1-Methylethoxy)ethoxy]methyl]phenoxy]-3-[(1-methylethyl)amino]-2-propanol hemifumarate salt
Empirical Formula (Hill Notation):
C18H31NO4 · .5C4H4O4
CAS Number:
Molecular Weight:
383.48

Products

Product Category
Brand
Color
Formula Weight
Gene Alias
Physical Form
Purity
Research Area
Showing 1-1 of 1
Compare
Product Number
Description
Pricing