![Biotinamidohexanoyl-6-aminohexanoic acid N-hydroxysuccinimide ester ≥95% (TLC), powder](/deepweb/assets/sigmaaldrich/product/images/369/837/c77276b2-1fa8-4b20-994f-00a62f84a83f/640/c77276b2-1fa8-4b20-994f-00a62f84a83f.jpg)
Biotinamidohexanoyl-6-aminohexanoic acid N-hydroxysuccinimide ester
![Biotinamidohexanoyl-6-aminohexanoic acid N-hydroxysuccinimide ester ≥95% (TLC), powder](/deepweb/assets/sigmaaldrich/product/images/369/837/c77276b2-1fa8-4b20-994f-00a62f84a83f/640/c77276b2-1fa8-4b20-994f-00a62f84a83f.jpg)
Synonym(s):
6-(Biotinamidocaproylamido)caproic acid N-hydroxysuccinimide ester, Biotin-X-X-NHS, N-Succinimidyl N-[6-(biotinylamino)caproyl]-6-aminocaproate
Empirical Formula (Hill Notation):
C26H41N5O7S
CAS Number:
Molecular Weight:
567.70
Products
Brand
Formula Weight
Physical Form
Purity
Quality Segment
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing