Skip to Content
Merck
Benzyldimethylhexylammonium chloride ≥96.0% (AT)

Benzyldimethylhexylammonium chloride

Benzyldimethylhexylammonium chloride ≥96.0% (AT)
Synonym(s):
Benzylhexyldimethylammonium chloride
Empirical Formula (Hill Notation):
C15H26ClN
CAS Number:
Molecular Weight:
255.83

Products

Brand
Formula Weight
Physical Form
Purity
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing