Skip to Content
Merck
Benzimidazole 98%

Benzimidazole

Benzimidazole 98%
Synonym(s):
1,3-Benzodiazole, BZI, Benzimidazole
Empirical Formula (Hill Notation):
C7H6N2
CAS Number:
Molecular Weight:
118.14

Products

Product Category
Brand
Formula Weight
Melting Point (°C)
Physical Form
Purity
Quality Segment
Markush Class
Markush Group
Showing 1-2 of 2
Compare
Product Number
Description
Pricing