Skip to Content
Merck
Benzenesulfonic acid monohydrate 97%

Benzenesulfonic acid monohydrate

Benzenesulfonic acid monohydrate 97%
Linear Formula:
C6H5SO3H · H2O
CAS Number:
Molecular Weight:
176.19
EC Number:

Products

Product Category
Brand
Formula Weight
Melting Point (°C)
Physical Form
Purity
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing