![Bafilomycin A1 from Streptomyces griseus ≥90% (HPLC)](/deepweb/assets/sigmaaldrich/product/structures/172/273/20ff8873-3d8d-4d72-8876-8dbe43b89de0/640/20ff8873-3d8d-4d72-8876-8dbe43b89de0.png)
Bafilomycin A1
![Bafilomycin A1 from Streptomyces griseus ≥90% (HPLC)](/deepweb/assets/sigmaaldrich/product/structures/172/273/20ff8873-3d8d-4d72-8876-8dbe43b89de0/640/20ff8873-3d8d-4d72-8876-8dbe43b89de0.png)
Synonym(s):
Bafilomycin A1, Streptomyces griseus, InSolution Bafilomycin A1
Empirical Formula (Hill Notation):
C35H58O9
CAS Number:
Molecular Weight:
622.83
Products
Product Category
Brand
Biological Source
Formula Weight
Physical Form
Purity
Activity Spectrum
Research Area
Quality Segment
Mode of Action
Showing 1-4 of 4
Compare
Product Number
Description
Pricing