![Aspirin Impurity D Pharmaceutical Secondary Standard; Certified Reference Material](/deepweb/assets/sigmaaldrich/product/images/189/432/11142f8f-d55a-4415-869b-a5b1400a7be9/640/11142f8f-d55a-4415-869b-a5b1400a7be9.jpg)
Aspirin Impurity D
![Aspirin Impurity D Pharmaceutical Secondary Standard; Certified Reference Material](/deepweb/assets/sigmaaldrich/product/images/189/432/11142f8f-d55a-4415-869b-a5b1400a7be9/640/11142f8f-d55a-4415-869b-a5b1400a7be9.jpg)
Synonym(s):
2-[[2-(Acetyloxy)benzoyl]oxy]benzoic acid, 2-Carboxyphenyl o-acetylsalicylate
Empirical Formula (Hill Notation):
C16H12O6
CAS Number:
Molecular Weight:
300.26
Products
Brand
Formula Weight
Agency
Format
Quality Segment
Application
Package Size
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing