Skip to Content
Merck
Asparagine monohydrate European Pharmacopoeia (EP) Reference Standard

Asparagine monohydrate

Asparagine monohydrate European Pharmacopoeia (EP) Reference Standard
Synonym(s):
L-Asparagine monohydrate, (S)-(+)-2-Aminosuccinamic acid, (S)-2-Aminosuccinic acid 4-amide, L-Aspartic acid 4-amide
Linear Formula:
NH2COCH2CH(NH2)COOH · H2O
CAS Number:
Molecular Weight:
150.13

Products

Formula Weight
Manufacturer
Melting Point (°C)
Format
Application
API Family
Reference Material Type
Special Grade