Skip to Content
Merck
Artesunate Pharmaceutical Secondary Standard; Certified Reference Material

Artesunate

Artesunate Pharmaceutical Secondary Standard; Certified Reference Material
Synonym(s):
Artesunate
Empirical Formula (Hill Notation):
C19H28O8
CAS Number:
Molecular Weight:
384.42

Products

Product Category
Brand
Biological Source
Color
Formula Weight
Manufacturer
Physical Form
Activity Spectrum
Agency
Format
Quality Segment
Application
Mode of Action
Package Size
API Family
Special Grade