
Arjunetin

Synonym(s):
(2α,3β,49α)- 2,3,19-Trihydroxyolean-12-en-28-oic acid β-D-glucopyranosyl ester, 24-Deoxysericoside
Empirical Formula (Hill Notation):
C36H58O10
CAS Number:
Molecular Weight:
650.84
Products
Formula Weight
Manufacturer
Physical Form
Product Line
Purity
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing