Skip to Content
Merck
Aristolochic acid II phyproof® Reference Substance

Aristolochic acid II

Aristolochic acid II phyproof® Reference Substance
Synonym(s):
Aristolochic acid B, 6-Nitrophenanthro(3,4-d)-1,3-dioxole-5-carboxylic acid, Aristolochic acid II, Noraristolochic acid
Empirical Formula (Hill Notation):
C16H9NO6
CAS Number:
Molecular Weight:
311.25

Products

Formula Weight
Manufacturer
Product Line
Format
Application
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing