Anti-Rabbit IgG (whole molecule), F(ab′)2 fragment−Alkaline Phosphatase antibody produced in goat
Products
Product Category
Brand
Biological Source
Clonality
Physical Form
Antibody Form
Research Area
Conjugate
Post Translational Modification
Species Reactivity
Technique
Showing 1-1 of 1