Skip to Content
Merck
Anti-DIS3 antibody produced in rabbit affinity isolated antibody, buffered aqueous glycerol solution

Anti-DIS3 antibody produced in rabbit

Anti-DIS3 antibody produced in rabbit affinity isolated antibody, buffered aqueous glycerol solution
Synonym(s):
Anti-Dis3 mitotic control homolog (S. cerevisiae), Anti-Dis3p, Anti-Exosc11, Anti-Exosome complex exonuclease RRP44, Anti-Kiaa1008, Anti-Rrp44, Anti-DIS3 mitotic control homolog (S. cerevisiae), Anti-DKFZp667L1817, Anti-FLJ10484, Anti-KIAA1008, Anti-MGC33035

Products

Product Category
Brand
Biological Source
Clonality
Feature
Gene Alias
Physical Form
Product Line
Antibody Form
Research Area
Conjugate
Quality Segment
Post Translational Modification
Species Reactivity
Technique